Merck
  • The Aurora kinase inhibitor SNS-314 shows broad therapeutic potential with chemotherapeutics and synergy with microtubule-targeted agents in a colon carcinoma model.
The Aurora kinase inhibitor SNS-314 shows broad therapeutic potential with chemotherapeutics and synergy with microtubule-targeted agents in a colon carcinoma model.
Molecular cancer therapeutics (2009-04-18)
Erica C VanderPorten, Pietro Taverna, Jennifer N Hogan, Marcus D Ballinger, W Michael Flanagan, Raymond V Fucini
ABSTRACT
Aurora kinases play key roles in regulating centrosome maturation, mitotic spindle formation, and cytokinesis during cell division, and are considered promising drug targets due to their frequent overexpression in a variety of human cancers. SNS-314 is a selective and potent pan Aurora inhibitor currently in a dose escalation phase 1 clinical trial for the treatment of patients with advanced solid tumors. Here, we report the antiproliferative effects of SNS-314 in combination with common chemotherapeutics in cell culture and xenograft models. The HCT116 colorectal carcinoma cell line, with intact or depleted p53 protein levels, was treated with SNS-314 and a cytotoxic chemotherapeutic from a panel comprised of gemcitabine, 5-fluorouracil (5-FU), carboplatin, daunomycin, SN-38 (the active metabolite of irinotecan), docetaxel, and vincristine. Combinations were administered under either concurrent or sequential schedules. SNS-314 has predominantly additive effects when administered concurrently with commonly used anticancer agents. Sequential administration of SNS-314 with chemotherapeutic compounds showed additive antiproliferative effects with carboplatin, gemcitabine, 5-FU, daunomycin, and SN-38, and synergy was observed in combination with gemcitabine, docetaxel, or vincristine. The most profound antiproliferative effects were observed with sequential administration of SNS-314 followed by docetaxel or vincristine. In vivo, SNS-314 potentiated the antitumor activity of docetaxel in xenografts. Both the in vitro synergies observed between SNS-314 and agents that target the mitotic spindle and the potentiation seen with docetaxel in vivo are consistent with a mechanism of action in which Aurora inhibition bypasses the mitotic spindle assembly checkpoint and prevents cytokinesis, augmenting subsequent spindle toxin-mediated mitotic catastrophe and cell death.
Sigma-Aldrich
Anti-phospho-Ser/Thr-Pro MPM-2 Antibody, clone MPM-2, Upstate®, from mouse
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder