Merck
  • Terlipressin: Current and emerging indications in chronic liver disease.
Terlipressin: Current and emerging indications in chronic liver disease.
Journal of gastroenterology and hepatology (2017-10-06)
Timothy Papaluca, Paul Gow
ABSTRACT
Terlipressin is an analogue of vasopressin that has potent vasoactive properties and has been available for use in most countries for nearly two decades. It has both established roles and emerging indications in the management of complications of decompensated chronic liver disease. We explore historic and emerging literature regarding the use of terlipressin for a range of indications including hepatorenal syndrome, portal hypertensive bleeding, and disruptions in sodium homeostasis. Novel methods of infusion-based terlipressin administration including the beneficial effect in reduction of adverse events are explored, in addition to new indications for the use of terlipressin in decompensated cirrhosis in an outpatient setting.