Merck
  • FAM19A1 is a new ligand for GPR1 that modulates neural stem-cell proliferation and differentiation.
FAM19A1 is a new ligand for GPR1 that modulates neural stem-cell proliferation and differentiation.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2018-05-26)
Can Zheng, Dixin Chen, Yan Zhang, Yun Bai, Shiyang Huang, Danfeng Zheng, Weiwei Liang, Shaoping She, Xinjian Peng, Pingzhang Wang, Xiaoning Mo, Quansheng Song, Ping Lv, Jing Huang, Richard D Ye, Ying Wang
ABSTRACT
FAM19A1 is a member of the family with sequence similarity 19 with unknown function. FAM19A1 mRNA expression is restricted to the CNS. Here, we report that FAM19A1 is a classic secretory protein, and expression levels correlate with brain development, increasing from embryonic d 12.5, peaking between postnatal d (P)1 and P7 and decreasing at wk 8. The adult hippocampus is a region of FAM19A1 high expression. Recombinant FAM19A1 suppressed the proliferation and self-renewal of neural stem cells (NSCs) and altered the lineage progression of NSCs with promoted neuron differentiation and suppressed astrocyte differentiation. Although GPCR 1 (GPR1) has been reported to be expressed in the CNS, its functions in the brain remain unclear. We identified GPR1 to be a functional receptor for FAM19A1. FAM19A1 interacted with GPR1 via the N-terminal domain (GPR1-ND), and its NSC modulatory functions required the Rho-associated protein kinase (ROCK) /ERK1/2 and ROCK/signal transducer and activator of transcription 3 signaling pathways. GPR1-ND that selectively bound to FAM19A1 neutralized the effects of FAM19A1 on NSC functions. Taken together, our results show, for the first time to our knowledge, that FAM19A1 is a novel regulatory factor of the proliferation and differentiation of NSCs, and identified a novel mechanism by which GPCR mediates the effects of FAM19A1 on NSC functions that may be important for brain development and neurogenesis. Additional exploration of the functions of FAM19A1 and GPR1 in the CNS may broaden the range of therapeutic options available for major brain disorders.-Zheng, C., Chen, D., Zhang, Y., Bai, Y., Huang, S., Zheng, D., Liang, W., She, S., Peng, X., Wang, P., Mo, X., Song, Q., Lv, P., Huang, J., Ye, R. D., Wang, Y. FAM19A1 is a new ligand for GPR1 that modulates neural stem-cell proliferation and differentiation.
Sigma-Aldrich
Mechlorethamine hydrochloride, 98%