Merck
  • The Relationship between RUVBL1 (Pontin, TIP49, NMP238) and BCL6 in Benign and Malignant Human Lymphoid Tissues.
The Relationship between RUVBL1 (Pontin, TIP49, NMP238) and BCL6 in Benign and Malignant Human Lymphoid Tissues.
Biochemistry and biophysics reports (2016-04-12)
Beverly W Baron, Rebecca M Baron, Joseph M Baron
ABSTRACT
The human BCL6 gene, which is involved in the pathogenesis of certain human lymphomas, encodes a transcriptional repressor that is needed for germinal center B cell development and T follicular helper cell differentiation. Our goal was to identify BCL6 target genes using a cell system in which BCL6 repressive effects are inhibited followed by subtractive hybridization, and we detected the RUVBL1 (Pontin, TIP49) gene as a potential target of BCL6 repression. Here we show that the BCL6 protein significantly represses RUVBL1 transcription (6.8-fold). Knockdown of endogenous BCL6 in a human B cell lymphoma line leads to significant upregulation of RUVBL1, and there is an inverse expression pattern between the BCL6 and RUVBL1 proteins in certain human lymphomas. RUVBL1 is part of the AAA+ superfamily and participates in multiple processes, including gene transcription regulation, chromatin remodeling, and DNA repair, which, if dysregulated, may promote lymphoma development. A further understanding of the relationship between RUVBL1 and BCL6 should improve our understanding of the pathogenesis of human lymphomas.
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution