Merck
  • Inhibition of TGF-β/Smad signaling by BAMBI blocks differentiation of human mesenchymal stem cells to carcinoma-associated fibroblasts and abolishes their protumor effects.
Inhibition of TGF-β/Smad signaling by BAMBI blocks differentiation of human mesenchymal stem cells to carcinoma-associated fibroblasts and abolishes their protumor effects.
Stem cells (Dayton, Ohio) (2012-10-05)
Lei Shangguan, Xinyu Ti, Ulf Krause, Bo Hai, Yanqiu Zhao, Zhenhua Yang, Fei Liu
ABSTRACT
Bone marrow mesenchymal stem cells (BM-MSCs) have multiple therapeutic potentials for regenerative, anti-inflammatory, and immunomodulatory purposes and also show promise as vehicles for gene therapy of various metastatic cancers based on their tumor-tropic capacity. However, BM-MSCs are also a source of carcinoma-associated fibroblasts (CAFs) and may promote growth and metastasis of cancer. Transforming growth factor β (TGF-β) signaling is required to induce CAF differentiation of mouse BM-MSCs in vivo and can induce expression of some CAF markers in human BM-MSCs in vitro. To determine whether inhibiting TGF-β signaling in human BM-MSCs can block their differentiation to CAFs induced by tumor microenvironments and the consequent protumor effects, we transduced human BM-MSCs with a lentiviral vector encoding bone morphogenetic protein and activin membrane-bound inhibitor (BAMBI), a decoy TGF-β receptor. BAMBI transduction significantly inhibited TGF-β/Smad signaling and expression of CAF markers in human BM-MSCs treated with TGF-β1 or tumor-conditioned medium or cocultured with cancer cells, but did not alter the stem cell properties and the tumor-tropic property of MSCs. In addition, BAMBI transduction disrupted the cytokine network mediating the interaction between MSCs and breast cancer cells. Consequently, BAMBI transduction abolished protumor effects of BM-MSCs in vitro and in an orthotopic breast cancer xenograft model, and instead significantly inhibited growth and metastasis of coinoculated cancer. These results indicated that TGF-β signaling is essential for differentiation of human BM-MSCs to CAFs in tumor microenvironments and the consequent protumor effects, and inhibiting TGF-β/Smad pathway may improve the safety of MSC-based therapies in cancer patients.
Sigma-Aldrich
Monoclonal Anti-Fibroblast Surface Protein antibody produced in mouse, clone 1B10, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Actin, α-Smooth Muscle, clone 1A4, ascites fluid