Merck
  • Light-Controlled Lipid Interaction and Membrane Organization in Photolipid Bilayer Vesicles.
Light-Controlled Lipid Interaction and Membrane Organization in Photolipid Bilayer Vesicles.
Langmuir : the ACS journal of surfaces and colloids (2018-10-23)
Patrick Urban, Stefanie D Pritzl, David B Konrad, James A Frank, Carla Pernpeintner, Christian R Roeske, Dirk Trauner, Theobald Lohmüller
ABSTRACT
Controlling lateral interactions between lipid molecules in a bilayer membrane to guide membrane organization and domain formation is a key factor for studying and emulating membrane functionality in synthetic biological systems. Here, we demonstrate an approach to reversibly control lipid organization, domain formation, and membrane stiffness of phospholipid bilayer membranes using the photoswitchable phospholipid azo-PC. azo-PC contains an azobenzene group in the sn2 acyl chain that undergoes reversible photoisomerization on illumination with UV-A and visible light. We demonstrate that the concentration of the photolipid molecules and also the assembly and disassembly of photolipids into lipid domains can be monitored by UV-vis spectroscopy because of a blue shift induced by photolipid aggregation.
Product Description
Avanti
4ME 16:0 PC, Avanti Research - A Croda Brand
Avanti
4ME 16:0 PC, Avanti Research - A Croda Brand