Merck
  • MicroRNA-499a-5p Promotes Differentiation of Human Bone Marrow-Derived Mesenchymal Stem Cells to Cardiomyocytes.
MicroRNA-499a-5p Promotes Differentiation of Human Bone Marrow-Derived Mesenchymal Stem Cells to Cardiomyocytes.
Applied biochemistry and biotechnology (2018-03-27)
Vajiheh Neshati, Samaneh Mollazadeh, Bibi Sedigheh Fazly Bazzaz, Antoine A F de Vries, Majid Mojarrad, Hojjat Naderi-Meshkin, Zeinab Neshati, Mahdi Mirahmadi, Mohammad Amin Kerachian
ABSTRACT
Since the adult mammalian heart has limited regenerative capacity, cardiac trauma, disease, and aging cause permanent loss of contractile tissue. This has fueled the development of stem cell-based strategies to provide the damaged heart with new cardiomyocytes. Bone marrow-derived mesenchymal stem cells (BM-MSCs) are capable of self-renewal and differentiation into cardiomyocytes, albeit inefficiently. MicroRNAs (miRNAs, miRs) are non-coding RNAs that have the potential to control stem cell fate decisions and are employed in cardiac regeneration and repair. In this study, we tested the hypothesis that overexpression of miR-499a induces cardiomyogenic differentiation in BM-MSCs. Human BM-MSCs (hBM-MSCs) were transduced with lentiviral vectors encoding miR-499a-3p or miR-499a-5p and analyzed by immunostaining and western blotting methods 14 days post-transduction. MiR-499a-5p-transduced cells adopted a polygonal/rod-shaped (myocyte-like) phenotype and showed an increase in the expression of the cardiomyocyte markers α-actinin and cTnI, as cardiogenic differentiation markers. These results indicate that miR-499a-5p overexpression promotes the cardiomyogenic differentiation of hBM-MSCs and may thereby increase their therapeutic efficiency in cardiac regeneration.