- Analogous structural motifs in myelin basic protein and in MARCKS.
Analogous structural motifs in myelin basic protein and in MARCKS.
Molecular and cellular biochemistry (2000-08-15)
G Harauz, N Ishiyama, I R Bates
PMID10942213
ABSTRACT
Myelin basic protein (MBP) and myristoylated alanine-rich C-kinase substrate (MARCKS) are similar in terms of having extended conformations regulated by their environment (i.e., solubilised or lipid-associated), N-terminal modifications, a dual nature of interactions with lipids, binding to actin and Ca2+-calmodulin, and being substrates for different kinds of protein kinases. The further sequence similarities of segments of MBP with lipid effector regions of MARCKS, and numerous reports in the literature, support the thesis that some developmental isoform of MBP functions in signal transduction.
Sigma-Aldrich
MBP, GST tagged human, recombinant, expressed in E. coli, ≥65% (SDS-PAGE), buffered aqueous glycerol solution