Merck
  • Regulation of lipoxygenase-1 and Dectin-1 on interleukin-10 in mouse Aspergillus fumigatus keratitis.
Regulation of lipoxygenase-1 and Dectin-1 on interleukin-10 in mouse Aspergillus fumigatus keratitis.
International journal of ophthalmology (2018-07-07)
Cheng-Ye Che, Ke-Lan Yuan, Gui-Qiu Zhao, Cui Li, Jing Lin, Guo-Qiang Zhu, Min Liu
ABSTRACT
To investigate the regulation of lipoxygenase (LOX)-1 and Dectin-1 on interleukin-10 (IL-10) production in mice with Aspergillus fumigatus (A. fumigatus) keratitis. The corneas of C57BL/6 mice were pretreated with LOX-1 inhibitor Poly(I) or Dectin-1 siRNA separately before the infection of A. fumigatus. Polymerase chain reaction (PCR) and Western blot were used to detect the expression of IL-10. The mRNA and protein expressions of IL-10 were significantly increased in mice with A. fumigatus keratitis. Compared with the group pretreated with sterile water before infection, Poly(I) pretreatment suppressed IL-10 expression significantly. Compared with the group pretreated with scrambled siRNA before infection, Dectin-1 siRNA pretreatment significantly reduced IL-10 expression in response to A. fumigatus infection. LOX-1 and Dectin-1 regulate IL-10 production in mouse A. fumigatus keratitis.