Merck
  • Design, synthesis, biological evaluation, and structure-activity relationships of substituted phenyl 4-(2-oxoimidazolidin-1-yl)benzenesulfonates as new tubulin inhibitors mimicking combretastatin A-4.
Design, synthesis, biological evaluation, and structure-activity relationships of substituted phenyl 4-(2-oxoimidazolidin-1-yl)benzenesulfonates as new tubulin inhibitors mimicking combretastatin A-4.
Journal of medicinal chemistry (2011-05-25)
Sébastien Fortin, Lianhu Wei, Emmanuel Moreau, Jacques Lacroix, Marie-France Côté, Eric Petitclerc, Lakshmi P Kotra, René C-Gaudreault
ABSTRACT
Sixty-one phenyl 4-(2-oxoimidazolidin-1-yl)benzenesulfonates (PIB-SOs) and 13 of their tetrahydro-2-oxopyrimidin-1(2H)-yl analogues (PPB-SOs) were prepared and biologically evaluated. The antiproliferative activities of PIB-SOs on 16 cancer cell lines are in the nanomolar range and unaffected in cancer cells resistant to colchicine, paclitaxel, and vinblastine or overexpressing the P-glycoprotein. None of the PPB-SOs exhibit significant antiproliferative activity. PIB-SOs block the cell cycle progression in the G(2)/M phase and bind to the colchicine-binding site on β-tubulin leading to cytoskeleton disruption and cell death. Chick chorioallantoic membrane tumor assays show that compounds 36, 44, and 45 efficiently block angiogenesis and tumor growth at least at similar levels as combretastatin A-4 (CA-4) and exhibit low to very low toxicity on the chick embryos. PIB-SOs were subjected to CoMFA and CoMSIA analyses to establish quantitative structure-activity relationships.
Sigma-Aldrich
Colchicine, ≥95% (HPLC), powder
Sigma-Aldrich
Colchicine, BioReagent, suitable for plant cell culture, ≥95% (HPLC)