Merck
  • Lin28 modulates cell growth and associates with a subset of cell cycle regulator mRNAs in mouse embryonic stem cells.
Lin28 modulates cell growth and associates with a subset of cell cycle regulator mRNAs in mouse embryonic stem cells.
RNA (New York, N.Y.) (2009-01-17)
Bingsen Xu, Kexiong Zhang, Yingqun Huang
ABSTRACT
Lin28 is highly expressed in human and mouse embryonic stem (ES) cells. Here, we show that in mouse ES cells, specific repression of Lin28 results in decreased cell proliferation, while overexpression of Lin28 accelerates cell proliferation. Further, Lin28 associates specifically with ribonucleoprotein particles containing mRNAs for cyclins A and B and cdk4. Importantly, changes in Lin28 levels lead to corresponding changes in the levels of these proteins, and sequences from the 3' untranslated regions of cyclin B and cdk4 mRNAs exhibit stimulatory effects on translation of reporter genes in a Lin28-dependent fashion. Thus, we postulate that Lin28 may play a role in the regulation of translation of genes important for the growth and maintenance of pluripotent cells.
Sigma-Aldrich
Lin28-TAT human, recombinant, expressed in E. coli, ≥90% (SDS-PAGE), ≥90% (HPLC), suitable for cell culture
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
IgG, Mouse
Sigma-Aldrich
Anti-Cyclin A Antibody, Upstate®, from rabbit