- Glutaredoxin 3 promotes migration and invasion via the Notch signalling pathway in oral squamous cell carcinoma.
Glutaredoxin 3 promotes migration and invasion via the Notch signalling pathway in oral squamous cell carcinoma.
Substantial evidence indicates that the alteration of the cellular redox status is a critical factor involved in cell growth and death and results in tumourigenesis. Cancer cells have an efficient antioxidant system to counteract the increased generation of ROS. However, whether this ability to survive high levels of ROS has an important role in the growth and metastasis of tumours is not well understood. Glutaredoxin 3 (GLRX3), also known as TXNL2, Grx3 and PICOT, maintains a low level of ROS, thus contributing to the survival and metastasis of several types of cancer. However, little is known about the role of GLRX3 and the underlying mechanisms that suppress oral squamous cell carcinoma (OSCC) progression. Here, by using immunohistochemical staining, we demonstrated that GLRX3 was overexpressed in human OSCC, and enhanced GLRX3 expression correlated with metastasis and with decreased overall patient survival. Knockdown of GLRX3 in human OSCC cell lines reduced Notch activity by reversing the epithelial-mesenchymal transition (EMT), resulting in the inhibition of in vitro migration and invasion. Importantly, knockdown of GLRX3 triggered the generation of ROS. Furthermore, N-acetyl cysteine (NAC), an ROS scavenger, enhanced the effects of GLRX3 knockdown on Notch-dependent EMT. Collectively, these findings suggested the vital roles of GLRX3 in OSCC progression through its relationship with EMT progression, and these data also suggest that a strategy of blocking ROS to enhance the activity of GLRX3 knockdown warrants further attention in the treatment of OSCC.