Merck
  • Increased B4GALT1 expression associates with adverse outcome in patients with non-metastatic clear cell renal cell carcinoma.
Increased B4GALT1 expression associates with adverse outcome in patients with non-metastatic clear cell renal cell carcinoma.
Oncotarget (2016-04-20)
Huyang Xie, Yu Zhu, Huimin An, Hongkai Wang, Yao Zhu, Hangcheng Fu, Zewei Wang, Qiang Fu, Jiejie Xu, Dingwei Ye
ABSTRACT
B4GALT1 is one of seven beta-1, 4-galactosyltransferase (B4GALT) genes, which has distinct functions in various malignances. Here, we evaluate the association of B4GALT1 expression with oncologic outcome in patients with non-metastatic clear cell renal cell carcinoma (ccRCC). A retrospective analysis of 438 patients with non-metastatic ccRCC at two academic medical centers between 2005 and 2009 was performed. The first cohort with 207 patients was treated as training cohort and the other as validation cohort. Tissue microarrays (TMAs) were created in triplicate from formalin-fixed, paraffin embedded specimens. Immunohistochemistry (IHC) was performed and the association of B4GALT1 expression with standard pathologic features and prognosis were evaluated. B4GALT1 expression was significantly associated with tumor T stage (P<0.001 and P<0.001, respectively), Fuhrman grade (P<0.001 and P<0.001, respectively) and necrosis (P=0.021 and P=0.002, respectively) in both training and validation cohorts. And high B4GALT1 expression indicated poor overall survival (OS) (P<0.001 and P<0.001, respectively) in the two cohorts. Furthermore, B4GALT1 expression was identified as an independent adverse prognostic factor for survival (P=0.007 and P=0.002, respectively). Moreover, the accuracy of established prognostic models was improved when B4GALT1 expression was added. Therefore, a predictive nomogram was generated with identified independent prognosticators to assess patients' OS at 5 and 10 years. Increased B4GALT1 expression is a potential independent adverse prognostic factor for OS in patients with non-metastatic ccRCC.