Merck
  • Overexpression of CD164 in oral cavity squamous cell carcinoma predicts a favourable prognosis.
Overexpression of CD164 in oral cavity squamous cell carcinoma predicts a favourable prognosis.
Oncology letters (2017-11-09)
Jia-Hong Chen, Wei-Liang Chen, James Yi-Hsin Chan, Yuan-Wu Chen, Yi-Jen Peng, Ming-Fang Cheng, Chun-Shu Lin
ABSTRACT
The aim of the present study was to investigate the association between cluster of differentiation (CD) 164 expression with clinicopathological parameters and prognosis among patients with oral cavity squamous cell carcinoma (OSCC). The present study retrospectively reviewed 70 patients with OSCC who underwent curative primary surgery. A number of patients subsequently received postoperative chemoradiotherapy although the specimens were not exposed to radiation or chemotherapy prior to anti-CD164 antibody immunohistochemical staining. CD164 overexpression was arbitrarily defined as exhibiting an H-score of ≥120. Univariate and multivariate analyses were performed for sex, age, American Joint Committee on Cancer stage, tumour location, histological grade, surgical margin and H-score. The 5-year overall survival rate was 54.4% and the median follow-up time was 46 months for surviving patients. Univariate analyses revealed that a low overall survival rate was associated with advanced-stage disease (P<0.001), buccogingival tumour location (P=0.038) and a CD164 H-score of <120 (P=0.016). Multivariate Cox's regression analyses revealed that poor overall survival rate was associated with advanced-stage disease (P=0.001) and a CD164 H-score of <120 (P=0.04). CD164 overexpression in OSCC was associated with favourable survival rate. Thus, CD164 expression may be a clinically useful predictor of prognosis in patients with OSCC.
Sigma-Aldrich
Monoclonal Anti-Rabbit Immunoglobulins–Biotin antibody produced in mouse, clone RG-16, purified immunoglobulin, buffered aqueous solution