![Xylose European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/201/314/5f94fe4c-fbfc-4e50-8e97-7999a3bebfb0/640/5f94fe4c-fbfc-4e50-8e97-7999a3bebfb0.png)
Xylose
![Xylose European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/201/314/5f94fe4c-fbfc-4e50-8e97-7999a3bebfb0/640/5f94fe4c-fbfc-4e50-8e97-7999a3bebfb0.png)
Synonym(s):
D-(+)-Xylose
Empirical Formula (Hill Notation):
C5H10O5
CAS Number:
Molecular Weight:
150.13
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing