![Vicenin 2 primary reference standard](/deepweb/assets/sigmaaldrich/product/structures/135/940/401c91d7-910c-4f3d-b558-9ae4d7e612dc/640/401c91d7-910c-4f3d-b558-9ae4d7e612dc.png)
Vicenin 2
![Vicenin 2 primary reference standard](/deepweb/assets/sigmaaldrich/product/structures/135/940/401c91d7-910c-4f3d-b558-9ae4d7e612dc/640/401c91d7-910c-4f3d-b558-9ae4d7e612dc.png)
Synonym(s):
6,8-Di-C-β-D-glucopyranosylapigenin, Apigenin 6,8-C-di-β-D-glucopyranoside, Apigenin 6,8-diglucoside
Empirical Formula (Hill Notation):
C27H30O15
CAS Number:
Molecular Weight:
594.52
Products
Formula Weight
Manufacturer
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing