![Valproic acid for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/105/661/21491845-8e0e-4bec-97f5-34056cb2bb4a/640/21491845-8e0e-4bec-97f5-34056cb2bb4a.png)
Valproic acid for system suitability
![Valproic acid for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/105/661/21491845-8e0e-4bec-97f5-34056cb2bb4a/640/21491845-8e0e-4bec-97f5-34056cb2bb4a.png)
Synonym(s):
2-Propylpentanoic acid, Valproic acid
Linear Formula:
(CH3CH2CH2)2CHCO2H
CAS Number:
Molecular Weight:
144.21
Products
Boiling Point (°C)
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing