Skip to Content
ID
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
UV curable resin
Products
Product Category
resins (4)
Brand
Sigma-Aldrich (4)
Feature
Physical Form
Quality Segment
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
900149
UV curable resin,
low viscosity
Expand
View Pricing
900166
UV curable resin,
adhesible to plastics and glass, low viscosity
Expand
View Pricing
900156
UV curable resin,
for protective coatings, optical, and outdoor applications
Expand
View Pricing
900164
UV curable resin,
rapid cure, adhesible to plastics and glass
Expand
View Pricing