Skip to Content
Merck
Tyrosinase from mushroom lyophilized powder, ≥1000 unit/mg solid

Tyrosinase from mushroom

Tyrosinase from mushroom lyophilized powder, ≥1000 unit/mg solid
Synonym(s):
Catechol Oxidase, Monophenol Monooxygenase, Monophenol, dihydroxyphenylalanine:oxygen oxidoreductase, Polyphenol Oxidase
CAS Number:

Products

Product Category
Brand
Biological Source
Physical Form
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing