![TWS119 ≥98% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/284/436/9780fb14-0416-4750-af85-fa800322f195/640/9780fb14-0416-4750-af85-fa800322f195.png)
TWS119
![TWS119 ≥98% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/284/436/9780fb14-0416-4750-af85-fa800322f195/640/9780fb14-0416-4750-af85-fa800322f195.png)
Synonym(s):
3-[6-(3-Aminophenyl)-7H-pyrrolo[2,3-d]pyrimidin-4-yloxy]phenol
Empirical Formula (Hill Notation):
C18H14N4O2
CAS Number:
Molecular Weight:
318.33
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing