
Trimethoprim for system suitability

Synonym(s):
Trimethoprim, 2,4-Diamino-5-(3,4,5-trimethoxybenzyl)pyrimidine, NSC 106568
Empirical Formula (Hill Notation):
C14H18N4O3
CAS Number:
Molecular Weight:
290.32
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing