Skip to Content
Merck
trans-9-Octadecenoic acid methyl ester certified reference material, 10 mg/mL in heptane, ampule of 1 mL

trans-9-Octadecenoic acid methyl ester

trans-9-Octadecenoic acid methyl ester certified reference material, 10 mg/mL in heptane, ampule of 1 mL
Synonym(s):
Methyl elaidate, Elaidic acid methyl ester, Methyl trans-9-octadecenoate
Linear Formula:
CH3(CH2)7CH=CH(CH2)7COOCH3
CAS Number:
Molecular Weight:
296.49

Products

Product Category
Brand
Formula Weight
Physical Form
Product Line
Format
Quality Segment
Functional Group
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing