![Tolazamide United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/270/944/3837c2f2-7543-43b1-a3bc-365e4bac4b7c/640/3837c2f2-7543-43b1-a3bc-365e4bac4b7c.png)
Tolazamide
![Tolazamide United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/270/944/3837c2f2-7543-43b1-a3bc-365e4bac4b7c/640/3837c2f2-7543-43b1-a3bc-365e4bac4b7c.png)
Synonym(s):
1-(Hexahydro-1H-azepin-1-yl)-3-(p-toluenesulfonyl)urea
Empirical Formula (Hill Notation):
C14H21N3O3S
CAS Number:
Molecular Weight:
311.40
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing