Skip to Content
Merck
Tissue Inhibitor of Metalloproteinase-3 human recombinant, expressed in NSO cells, >95% (SDS-PAGE)

Tissue Inhibitor of Metalloproteinase-3 human

Tissue Inhibitor of Metalloproteinase-3 human recombinant, expressed in NSO cells, >95% (SDS-PAGE)
Synonym(s):
TIMP-3

Products

Product Category
Brand
Biological Source
Gene Alias
Physical Form
Purity
Recombinant Host
Quality Segment
Technique
Showing 1-1 of 1