![Thyrotropic hormone from human pituitary lyophilized powder, ≥6.2 IU/mg (immunoassay, non-sterile)](/deepweb/assets/sigmaaldrich/product/images/157/247/ed9836ed-f6fb-4bda-96eb-232442186541/640/ed9836ed-f6fb-4bda-96eb-232442186541.jpg)
Thyrotropic hormone from human pituitary
![Thyrotropic hormone from human pituitary lyophilized powder, ≥6.2 IU/mg (immunoassay, non-sterile)](/deepweb/assets/sigmaaldrich/product/images/157/247/ed9836ed-f6fb-4bda-96eb-232442186541/640/ed9836ed-f6fb-4bda-96eb-232442186541.jpg)
Synonym(s):
TSH, Thyroid stimulating hormone
CAS Number:
Products
Product Category
Brand
Biological Source
Gene Alias
Physical Form
Tissue
Quality Segment
Sterility
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing