
Thioridazine for system suitability

Synonym(s):
Thioridazine, 10-[2-(1-Methyl-2-piperidinyl)ethyl]-2-(methylthio)-10H-phenothiazine
Empirical Formula (Hill Notation):
C21H26N2S2
CAS Number:
Molecular Weight:
370.57
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing