![Tetrakis(dimethylamido)tin(IV) 99.9% trace metals basis](/deepweb/assets/sigmaaldrich/product/structures/180/137/10108b70-e950-412c-87f2-7dd79d1707c9/640/10108b70-e950-412c-87f2-7dd79d1707c9.png)
Tetrakis(dimethylamido)tin(IV)
![Tetrakis(dimethylamido)tin(IV) 99.9% trace metals basis](/deepweb/assets/sigmaaldrich/product/structures/180/137/10108b70-e950-412c-87f2-7dd79d1707c9/640/10108b70-e950-412c-87f2-7dd79d1707c9.png)
Synonym(s):
Tin(IV) dimethylamide
Linear Formula:
[(CH3)2N]4Sn
CAS Number:
Molecular Weight:
295.01
Products
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing