
Synthechol® Synthetic Cholesterol

Synonym(s):
Cholesterol, 3β-Hydroxy-5-cholestene, 5-Cholesten-3β-ol
Empirical Formula (Hill Notation):
C27H46O
CAS Number:
Molecular Weight:
386.65
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Agency
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing