
Succinimide

Synonym(s):
2,5-Pyrrolidinedione, Succinimide
Empirical Formula (Hill Notation):
C4H5NO2
CAS Number:
Molecular Weight:
99.09
Products
Product Category
Brand
Boiling Point (°C)
Color
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Showing 1-2 of 2