![S-Phenyl-L-cysteine 97%](/deepweb/assets/sigmaaldrich/product/structures/410/612/50be19ff-3c56-44a1-945f-583c90408fca/640/50be19ff-3c56-44a1-945f-583c90408fca.png)
S-Phenyl-L-cysteine
![S-Phenyl-L-cysteine 97%](/deepweb/assets/sigmaaldrich/product/structures/410/612/50be19ff-3c56-44a1-945f-583c90408fca/640/50be19ff-3c56-44a1-945f-583c90408fca.png)
Synonym(s):
3-(Phenylthio)-L-Alanine, 4-Thia-L-homophenylalanine
Linear Formula:
C6H5SCH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
197.25
Products
Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing