
Solanidine

Synonym(s):
(3β)-Solanid-5-en-3-ol, Solatubine
Empirical Formula (Hill Notation):
C27H43NO
CAS Number:
Molecular Weight:
397.64
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing