Skip to Content
Merck
Sodium formate-13C,18O2 95 atom % 18O, 99 atom % 13C

Sodium formate-13C,18O2

Sodium formate-13C,18O2 95 atom % 18O, 99 atom % 13C
Synonym(s):
Formic acid-13C,18O2 sodium salt
Linear Formula:
H13C18O2Na
CAS Number:
Molecular Weight:
73.00

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Isotope
Showing 1-1 of 1
Compare
Product Number
Description
Pricing