Rosuvastatin for system suitability
Synonym(s):
Rosuvastatin calcium salt
Empirical Formula (Hill Notation):
C44H54CaF2N6O12S2
CAS Number:
Molecular Weight:
1001.14
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing