RDX solution
Synonym(s):
Hexahydro-1,3,5-trinitro-1,3,5-triazine solution
Empirical Formula (Hill Notation):
C3H6N6O6
CAS Number:
Molecular Weight:
222.12
Products
Brand
Formula Weight
Manufacturer
Format
Application
Package Size
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing