Pyraclostrobin
Synonym(s):
Methyl {2-[1-(4-chlorophenyl)-1H-pyrazol-3-yloxymethyl]phenyl}methoxycarbamate
Empirical Formula (Hill Notation):
C19H18ClN3O4
CAS Number:
Molecular Weight:
387.82
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2