Skip to Content
Merck
Procyanidin B2 analytical standard

Procyanidin B2

Procyanidin B2 analytical standard
Synonym(s):
(−)-Epicatechin (4β-8)-(−)-epicatechin, 4,8″-Bi-[(+)-epicatechin], cis,cis″-4,8″-Bi(3,3′,4′,5,7-pentahydroxyflavane), Proanthocyanidin B2
Empirical Formula (Hill Notation):
C30H26O12
CAS Number:
Molecular Weight:
578.52

Products

Brand
Formula Weight
Manufacturer
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing