Skip to Content
Merck
Primuline Dye content 50 %

Primuline

Primuline Dye content 50 %
Synonym(s):
Direct Yellow 59, Primuline Yellow
Empirical Formula (Hill Notation):
C21H14N3NaO3S3
CAS Number:
Molecular Weight:
475.54

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Chemical Composition
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing