Skip to Content
Merck
Prednisolone hemisuccinate United States Pharmacopeia (USP) Reference Standard

Prednisolone hemisuccinate

Prednisolone hemisuccinate United States Pharmacopeia (USP) Reference Standard
Synonym(s):
Prednisolone 21-hemisuccinate, Δ1-Hydrocortisone 21-hemisuccinate, (11β)-11,17-Dihydroxy-21-[(3,3-dimethyl-1-oxobutyl)oxy]pregna-1,4-diene-3,20-dione, 1,4-Pregnadiene-11β,17,21-triol-3,20-dione 21-hemisuccinate, 11β,17α,21-Trihydroxy-1,4-pregnadiene-3,20-dione 21-hemisuccinate
Empirical Formula (Hill Notation):
C25H32O8
CAS Number:
Molecular Weight:
460.52
EC Number:

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing