
Plastic additive 1

Synonym(s):
Ethylene bis[3,3-bis[3-(1,1-dimethylethyl)-4-hydroxyphenyl]butanoate]
Empirical Formula (Hill Notation):
C50H66O8
CAS Number:
Molecular Weight:
795.05
Products
Formula Weight
Manufacturer
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing