Skip to Content
Merck
Phosphatase substrate Suitable for manufacturing of diagnostic kits and reagents

Phosphatase substrate

Phosphatase substrate Suitable for manufacturing of diagnostic kits and reagents
Synonym(s):
4-Nitrophenyl phosphate disodium salt hexahydrate, pNPP disodium salt hexahydrate, Phosphatase substrate
Linear Formula:
O2NC6H4OP(O)(ONa)2 · 6H2O
CAS Number:
Molecular Weight:
371.14
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Application