Skip to Content
Merck
Phloretin ≥99%

Phloretin

Phloretin ≥99%
Synonym(s):
β-(4-Hydroxyphenyl)-2,4,6-trihydroxypropiophenone, 2′,4′,6′-Trihydroxy-3-(4-hydroxyphenyl)propiophenone, 3-(4-Hydroxyphenyl)-1-(2,4,6-trihydroxyphenyl)-1-propanone
Empirical Formula (Hill Notation):
C15H14O5
CAS Number:
Molecular Weight:
274.27
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing