Skip to Content
Merck
ω-Pentadecalactone ≥98%, FG

ω-Pentadecalactone

ω-Pentadecalactone ≥98%, FG
Synonym(s):
Pentadecanolide, 1-Oxa-2-cyclohexadecanone, 15-Hydroxypentadecanoic acid lactone, 15-Pentadecanolide, Pentalide
Empirical Formula (Hill Notation):
C15H28O2
CAS Number:
Molecular Weight:
240.38
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing