![N-Succinyl-Leu-Tyr-7-amido-4-methylcoumarin calpain substrate](/deepweb/assets/sigmaaldrich/product/structures/761/217/76de81f9-1c9a-4844-9df8-4f11fcb6c0bf/640/76de81f9-1c9a-4844-9df8-4f11fcb6c0bf.png)
N-Succinyl-Leu-Tyr-7-amido-4-methylcoumarin
![N-Succinyl-Leu-Tyr-7-amido-4-methylcoumarin calpain substrate](/deepweb/assets/sigmaaldrich/product/structures/761/217/76de81f9-1c9a-4844-9df8-4f11fcb6c0bf/640/76de81f9-1c9a-4844-9df8-4f11fcb6c0bf.png)
Empirical Formula (Hill Notation):
C29H33N3O8
CAS Number:
Molecular Weight:
551.59
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing