![Noradrenaline tartrate European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/228/284/f06f53ee-775e-4829-89b9-710c1b2be7a7/640/f06f53ee-775e-4829-89b9-710c1b2be7a7.png)
Noradrenaline tartrate
![Noradrenaline tartrate European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/228/284/f06f53ee-775e-4829-89b9-710c1b2be7a7/640/f06f53ee-775e-4829-89b9-710c1b2be7a7.png)
Synonym(s):
L-(−)-Norepinephrine (+)-bitartrate salt monohydrate, L-(−)-Noradrenaline (+)-bitartrate salt monohydrate
Empirical Formula (Hill Notation):
C8H11NO3 · C4H6O6 · H2O
CAS Number:
Molecular Weight:
337.28
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing