Skip to Content
Merck
Nodakenin phyproof® Reference Substance

Nodakenin

Nodakenin phyproof® Reference Substance
Synonym(s):
(+)-Marmesinin, (+)-Mermesinin, Nodakenetin β-D-glucopyranoside
Empirical Formula (Hill Notation):
C20H24O9
CAS Number:
Molecular Weight:
408.40

Products

Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing