Skip to Content
Merck
N,N′-Ditridecylperylene-3,4,9,10-tetracarboxylic diimide 95%

N,N′-Ditridecylperylene-3,4,9,10-tetracarboxylic diimide

N,N′-Ditridecylperylene-3,4,9,10-tetracarboxylic diimide 95%
Empirical Formula (Hill Notation):
C50H62N2O4
CAS Number:
Molecular Weight:
755.04

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing