Skip to Content
Merck
Nitrogen dioxide ≥99.5%

Nitrogen dioxide

Nitrogen dioxide ≥99.5%
Synonym(s):
Dinitrogen tetroxide, Nitrogen peroxide, Nitrogen tetroxide
Empirical Formula (Hill Notation):
NO2
CAS Number:
Molecular Weight:
46.01
EC Number:

Products

Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing