![Niaproof® 4 ~27% in H2O](/deepweb/assets/sigmaaldrich/product/structures/294/414/e21a27d8-2dda-4e9a-ac3d-5f0342b1411b/640/e21a27d8-2dda-4e9a-ac3d-5f0342b1411b.png)
Niaproof® 4
![Niaproof® 4 ~27% in H2O](/deepweb/assets/sigmaaldrich/product/structures/294/414/e21a27d8-2dda-4e9a-ac3d-5f0342b1411b/640/e21a27d8-2dda-4e9a-ac3d-5f0342b1411b.png)
Synonym(s):
7-Ethyl-2-methyl-4-undecyl sulfate sodium salt, Sodium 7-ethyl-2-methyl-4-undecyl sulfate, Tergitol™ 4
Empirical Formula (Hill Notation):
C14H29NaO4S
CAS Number:
Molecular Weight:
316.43
Products
Product Category
Brand
Formula Weight
Physical Form
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing