N-Cinnamoyl-N-debenzoylpaclitaxel
Empirical Formula (Hill Notation):
C49H53NO14
CAS Number:
Molecular Weight:
879.94
Products
Brand
Formula Weight
Manufacturer
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing