![N-(2-Hydroxyethyl)maleimide 97%](/deepweb/assets/sigmaaldrich/product/structures/395/964/352833ac-8c02-4258-9b84-b346184e6201/640/352833ac-8c02-4258-9b84-b346184e6201.png)
N-(2-Hydroxyethyl)maleimide
![N-(2-Hydroxyethyl)maleimide 97%](/deepweb/assets/sigmaaldrich/product/structures/395/964/352833ac-8c02-4258-9b84-b346184e6201/640/352833ac-8c02-4258-9b84-b346184e6201.png)
Synonym(s):
1-(2-Hydroxyethyl)-pyrrole-2,5-dione, 1-(2-Hydroxyethyl)maleimide, N-(Ethanol)maleimide, HEMI
Empirical Formula (Hill Notation):
C6H7NO3
CAS Number:
Molecular Weight:
141.12
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing